Learn More
Invitrogen™ REA Polyclonal Antibody

Description
Recommended positive controls: H1299, HeLa, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (87%), Pig (98%), Chicken (95%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | REA |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PHB2 |
| Gene Accession No. | O35129, Q5XIH7, Q99623 |
| Gene Alias | AU044498; BAP; Bap37; Bap-37; Bcap27; BCAP37; B-cell associated protein; B-cell receptor associated protein 37; B-cell receptor-associated protein 37; B-cell receptor-associated protein BAP37; D-prohibitin; hBAP; MGC117268; p22; phb2; phb2 {ECO:0000250; PNAS-141; prohibitin 2; prohibitin-2; Rea; Repressor of estrogen receptor activity; UniProtKB:Q99623} |
| Gene Symbols | PHB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.