Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581532
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA581532 100 μL
1 options

Catalog No. PIPA581532

Supplier: Invitrogen™ PA581532

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human REA/PHB2.

PHB2 acts as a mediator of transcriptional repression by nuclear hormone receptors via recruitment of histone deacetylases. This protein functions as an estrogen receptor (ER)-selective coregulator that potentiates the inhibitory activities of antiestrogens and represses the activity of estrogens. It competes with NCOA1 for modulation of ER transcriptional activity. It is probably involved in regulating mitochondrial respiration activity and in aging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REA
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PHB2
Gene Accession No. Q99623
Gene Alias AU044498; BAP; Bap37; Bap-37; Bcap27; BCAP37; B-cell associated protein; B-cell receptor associated protein 37; B-cell receptor-associated protein 37; B-cell receptor-associated protein BAP37; D-prohibitin; hBAP; MGC117268; p22; phb2; phb2 {ECO:0000250; PNAS-141; prohibitin 2; prohibitin-2; Rea; Repressor of estrogen receptor activity; UniProtKB:Q99623}
Gene Symbols PHB2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Human PHB2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11331
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.