Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Reactive Blue 4, dye content min. 35%

Catalog No. aaa1291114
Click to view available options
Quantity:
25 g
100 g

CAS: 13324-20-4 | C23H14Cl2N6O8S2 | 637.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13324-20-4
Molecular Formula C23H14Cl2N6O8S2
Molecular Weight (g/mol) 637.42
MDL Number MFCD00001218
InChI Key RTLULCVBFCRQKI-UHFFFAOYSA-N
Synonym C.I. 61205; Procion™ Blue MX-R
PubChem CID 25863
IUPAC Name 1-amino-4-[3-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]-4-sulfoanilino]-9,10-dioxoanthracene-2-sulfonic acid
SMILES NC1=C(C=C(NC2=CC=C(C(NC3=NC(Cl)=NC(Cl)=N3)=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O

Specifications

Formula Weight 637.44
Odor Odorless
Quantity 25 g
Chemical Name or Material Reactive Blue 4, dye content ≈40%
Hazard Category H314-H317-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
EINECSNumber 236-363-9
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.