Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ready-to-Assay™ Metabotropic Glutamate Receptor Frozen Cells

Catalog No. hts146rta Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 Kit

Used in Calcium Flux FLIPR Assays

  • Human mGlu2 GPCR frozen cells
  • Simple, rapid calcium assays with no requirement for intensive cell culturing
  • High viability functionality post-thaw
  • Detection method: Fluorescent
  • Host Cells: Chem-1
  • Protein Target: mGlu2
  • Target Sub-Family: Glutamate (Metabotropic)
  • Cell Line Type: GPCR Cell Lines
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.