Learn More
Cytiva Ready-To-Go™ T4 DNA Ligase
Catalog No. 45001317 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 Reactions
Catalog No. Quantity
45-001-317 50 Reactions
1 options
Catalog No. 45-001-317 Supplier Cytiva Supplier No. 27036101
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
For covalently linking DNA fragments in as little as 30 minutes
  • Catalyzes the formation of a phosphodiester bond between the 5'-phosphoryl group and the 3'-hydroxyl group of two double-stranded DNA fragments
  • Consists of a glassified reaction mix predispensed in single-reaction tubes
  • When reconstituted to final reaction volume of 20μL, yields solution of reaction buffer, T4 DNA ligase, and ATP
TRUSTED_SUSTAINABILITY
Specifications
Components FPLCpure™ T4 DNA Ligase 6 weiss units, 66mM Tris HCl (pH 7.6), 6.6mM MgCl2, 0.1mM ATP, 0.1mM Spermidine, 10mM DTT and stabilizer, glassified reaction mix predispensed in single-reaction tubes
For Use With (Application) Self-circularization of linear DNA prior to transformation, ligation of insert to vector and ligation of synthetic linkers to blunt-ended DNA
Storage Buffer TE Buffer (10mM Tris HCl, pH 8, with 1mM EDTA) or 1-2X One-phor-all Buffer
Content And Storage Room Temperature
Quantity 50 Reactions
Product Type Ready-To-Go T4 DNA Ligase, 50 reaction beads