Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reagent 2, Reagents
Reagent 2, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CR0561001B
| Quantity | 1 L |
|---|
Chemical Identifiers
| 7732-18-5, 67-56-1, 89-25-8 | |
| 18.02 | |
| water |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| O |
Specifications
| Pale Purple/Brown | |
| H2O | |
| O | |
| 18.02 | |
| Opaque HDPE | |
| 1 L | |
| Liquid |
| 7732-18-5, 67-56-1, 89-25-8 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.0187 | |
| Non-sterile | |
| Reagent 2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction