Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reagent 2, Reagents
Reagent 2, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$87.10 - $463.50
Chemical Identifiers
CAS | 7732-18-5, 67-56-1, 89-25-8 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-334-071
|
Reagents Holdings Llc
CR05610010F |
10 L |
Each for $463.50
|
|
|||||
01-334-070
|
Reagents Holdings Llc
CR0561001B |
1 L |
Each for $87.10
|
|
|||||
Chemical Identifiers
7732-18-5, 67-56-1, 89-25-8 | |
18.02 | |
water |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
Specifications
Pale Purple/Brown | |
H2O | |
O | |
18.02 | |
Cubitainer | |
Non-sterile | |
Reagent 2 |
7732-18-5, 67-56-1, 89-25-8 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1.0187 | |
Liquid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title