Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reagent 3, DNTB for Sulfite Analysis, Reagents
Reagent 3, DNTB for Sulfite Analysis, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CR0008004A
Quantity | 4 L |
---|
Chemical Identifiers
7732-18-5, 64-17-5, 69-78-3 | |
18.02 | |
water |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
Specifications
Colorless to Light Yellow | |
100°C | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1.0035 | |
Non-sterile | |
Reagent 3, DNTB for Sulfite Analysis |
7732-18-5, 64-17-5, 69-78-3 | |
H2O | |
O | |
18.02 | |
Poly Bottle | |
4 L | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction