Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent 3, DNTB for Sulfite Analysis, Reagents

Reagent 3, DNTB for Sulfite Analysis, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$1471.00 - $3680.00

Chemical Identifiers

CAS 7732-18-5, 64-17-5, 69-78-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-334-078
View Documents
Reagents Holdings Llc
CR00080010F
10 L
Each for $3,680.00
Only null left
Add to Cart
 
01-334-077
View Documents
Reagents Holdings Llc
CR0008004A
4 L
Each for $1,471.00
Only null left
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5, 64-17-5, 69-78-3
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Colorless to Light Yellow
100°C
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.0035
Liquid
7732-18-5, 64-17-5, 69-78-3
H2O
O
18.02
Cubitainer
Non-sterile
Reagent 3, DNTB for Sulfite Analysis
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.