Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol (Denatured Alcohol), 18.5% (v/v), Ricca Chemical
SDP

Catalog No. r2546185120 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
Packaging:
Natural Poly Bottle

Catalog No. R2546185120

Supplier: Ricca Chemical Company R2546185120A

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5, 64-17-5, 67-63-0, 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) Mixture
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Boiling Point 100°C
Quantity 120 mL
Packaging Natural Poly Bottle
Assay Percent Range 18.5% (v/v)
Percent Purity 18.5%
Grade Laboratory
Chemical Name or Material Reagent Alcohol, Denatured Alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.