Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ Reagent Alcohol, 70%
GSA_VASDP

Catalog No. stl70701
Click to view available options
Quantity:
4 x 1 gal.
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.

A high quality dehydrant to ensure optimal staining.

  • 100% reagent alcohol with 30 parts Type 1 water
  • Composition: Ethyl alcohol (56-70%), Isopropyl alcohol (0.7-10.5%), Methanol (0.7-10.5%)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Dilution 100% reagent alcohol with 30 parts Type 1 water
Concentration 0.7
Grade ACS
Quantity 4 x 1 gal.
Format Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.