Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol), Reagents

Catalog No. 01338143
Click to view available options
Quantity:
500 mL
1 L
4 L
200 L
Packaging:
Glass Bottle
Poly Bottle
Poly Drum
Tin Can

Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5, 67-63-0, 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 72°C
Quantity 4 L
Packaging Tin Can
Physical Form Liquid
Specific Gravity 0.785
Percent Purity Min. 99.5%
Grade ACS
Chemical Name or Material Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.