Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ricca Chemical Company Reagent Alcohol (Denatured Alcohol), 70% (v/v), Ricca Chemical
SDP

Catalog No. R254670038 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
3.8 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R254670038 3.8 L Natural Poly Bottle
25467032 1 L Natural Poly Bottle
25-467-01SC 4 L Safety-coated Glass Bottle
25467025 10 L Cubitainer
2546705 20 L Cubitainer
25-467-01 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R254670038 Supplier Ricca Chemical Company Supplier No. R25467003.8A
Add to Cart
Add to Cart
  • Reagent Alcohol (Denatured Alcohol), 70% (v/v)

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-63-0 , 67-56-1 , 67-56-1
Molecular Weight (g/mol) Mixture

Specifications

Melting Point Approximately 0°C
Color Colorless
Quantity 3.8 L
Packaging Natural Poly Bottle
Molecular Formula C3H8O
Synonym Ethyl Alcohol
Boiling Point Approximately 93°C
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Percent Purity 70%
Grade Laboratory
Chemical Name or Material Alcohol, Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.