Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beckman Coulter™ Reagent Bottle Caps
Shop All Beckman Coulter Products
Click to view available options
For Use With (Equipment):
Beckman Coulter 30 mL Bottles
Beckman Coulter 60 mL Bottles
Description
- For use with Beckman Coulter analyzers

Specifications
Specifications
For Use With (Equipment) | Beckman Coulter 30 mL Bottles |
Quantity | 6/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction