Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Reagent Diluent Concentrate 2
GSA_VASDP

Catalog No. DY995 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 x 21 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY995 5 x 21 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY995 Supplier R&D Systems™ Supplier No. DY995

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

10X Concentrated 10% BSA Solution for DuoSets

  • 10% BSA solution in a concentrated formula which can be diluted to make a working solution at a range of final BSA concentrations suitable for the assay being run
  • Make all solutions fresh or filter sterilize and store for up to 1 week at 2° to 8°C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

For Use With (Application) ELISA
Product Line R&D Systems
Quantity 5 x 21 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.