Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Reagent Refill for Digital Topsoil Tour

Shop All LaMotte Company Products


Click to view available options
Class Size:
50 Students
Description
- Set of consumable reagents as supplied in the original kit
Specifications
Specifications
Product Type | Topsoil Tour Reagent Refill |
Grade | K to 12 |
Includes | Replacement Reagents |
Class Size | 50 Students |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction