Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Reagent Refills
SDPSCI_ED

Catalog No. s72386 Shop All LaMotte Company Products
Click to view available options
Type:
Alkalinity Reagent B
Ammonia Reagent 1
Chlorine Reagent 1
Chlorine Reagent 3
Chlorine Reagent Refill
DPD 1 Powder
Ferroin Indicator
Mixed Acid
Nitrogen Reducing Reagent
Potassium Iodide
Sulfide 1
Sulfide 2
Sulfide Test 3
Sulfide Test 4
Volume (English):
0.01 lb.
0.5 oz.
1.01 oz.
2.02 oz.
0.22 lb.
16.9 oz.
32.1 oz.

Catalog No. S72386

Supplier: LaMotte™ 6807J

Only null left
Add to Cart
Add to Cart

Testing supplies for the discerning scientist

Refills for sulfide tests, potassium iodide, chlorine, ferroine indicators and more

Specifications

Storage Requirements 25°C
Type DPD 1 Powder
Volume (English) 0.22 lb.
Weight (Metric) 100 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.