Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Reagent Reservoir
Shop All Fisherbrand Products

Click to view available options
Capacity (Metric):
5 mL
10 mL
Sterility:
Non-sterile
Sterile
Packaging:
Bulk Wrapped
Individually Wrapped
Description
- Superior durability with enhanced walls and crush-resistant 'turrets' at the corners
- Easy handling with tabs to prevent stacked reservoirs from sticking together
- Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces wastage of expensive reagents
- Pour-off Spouts conveniently located on all four corners enables easy spill-free pouring and reduces chance of dripping
- Divided reservoir design allows pipetting of two different reagents or maximizes recovery if using only single channel and small amount of reagent
- Manufactured and gamma irradiated in an ISO certified facility
- Certified DNase, RNase and pyrogen free
- Packaged individually in an easy open HDPE package
Specifications
Specifications
Capacity (English) | 0.17 oz. |
Certifications/Compliance | DNase, RNase, and Pyrogen free quality certificate available upon request, Traceability Certificate available upon request |
Color | White |
Capacity (Metric) | 5 mL |
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir |
For Use With (Application) | Pipetting procedures |
Packaging | Individually Wrapped |
Quantity | 50 Pk. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction