Learn More
Fisherbrand™ Reagent Reservoir Chiller, Pack of 2
Description
- Reagents are kept at less than 5 °C (41 °F) for approximately 1 hour.
- Engineered custom fit design for Heathrow Scientific 50 or 100 mL reservoirs to ensure a maximum contact for longer uniform cooling.
- Eliminates the risk of contamination from messy ice bath.
- Not for use with liquid nitrogen.
- Symmetrical cooler design allows reagent reservoirs to be mounted in either direction.
- Stackable, saves storage and work space.
- 2-Year Warranty.
Specifications
Specifications
| Color | Purple |
| Capacity (Metric) | 50/55 mL |
| Dimensions (L x W x H) | 6.1 x 2.9 x 1.7 in. |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Reservoir Cooler |
| Quantity | 2 |
| Autoclavable | Not Autoclavable |
| Channels | 1 |
| Disposable | Reusable |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.