Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Reagent Reservoir Chiller, Pack of 2

Catalog No. 14127073
Change view
Click to view available options
Capacity (Metric):
100 mL
50/55 mL
Color:
Blue
Purple
Purple, Blue
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Color
14-127-073 50/55 mL Blue
14-127-059 50/55 mL Purple
14-127-068 50/55 mL Purple, Blue
14-127-069 100 mL Purple
14-127-070 100 mL Blue
14-127-071 100 mL Purple, Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-127-073 Supplier Fisherbrand™ Supplier No. 120731F2
Only null left
Add to Cart
Add to Cart

Reliable cooling, no ice required

  • Reagents are kept at less than 5 °C (41 °F) for approximately 1 hour.
  • Engineered custom fit design for Heathrow Scientific 50 or 100 mL reservoirs to ensure a maximum contact for longer uniform cooling.
  • Eliminates the risk of contamination from messy ice bath.
  • Not for use with liquid nitrogen.
  • Symmetrical cooler design allows reagent reservoirs to be mounted in either direction.
  • Stackable, saves storage and work space.
  • 2-Year Warranty.

Specifications

Color Blue
Capacity (Metric) 50/55 mL
Dimensions (L x W x H) 6.1 x 2.9 x 1.7 in.
Material Polypropylene
Sterility Non-sterile
Type Reservoir Cooler
Quantity 2
Autoclavable Not Autoclavable
Channels 1
Disposable Reusable
Bottom Shape V-bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.