Learn More
Fisherbrand™ Reagent Reservoir, Single wrap
Description
- Superior durability with enhanced walls and crush-resistant turrets at the corners
- Easy handling with tabs to prevent stacked reservoirs from sticking together
- Manufactured and gamma irradiated in an ISO certified facility
- Certified DNase RNase and pyrogen free
- Packaged individually in an easy open HDPE package
- Traceability Certificate available upon request
Specifications
Specifications
| Capacity (Metric) | 55 mL |
| Dimensions (L x W x H) | 4.5 x 1.1 x 1.4 in. |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reagent Reservoir |
| Packaging | 100/Bag |
| Quantity | 100 |
| Autoclavable | Not Autoclavable |
| Channels | 1 |
| Disposable | Single-use |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.