Learn More
YSI™ Reagent Starter Kits for 9300 and 9500 Photometers
Shop All Yellow Springs Instrument (YSI) ProductsDescription
- Includes reagents sufficient for 50 tests
Specifications
Specifications
| Type | Magnesium |
| For Use With (Equipment) | 9300 and 9500 Photometer |
| Includes | Reagents, instructions, accessories |
| Packaging | 50 Tests/Pk. |
| Range | 0 to 100 mg/L |