Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Reagent Starter Kits for 9300 and 9500 Photometers

Catalog No. 1539929 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Type:
Alkalinity, Total
Ammonia
Calcium Hardness
Chlorine-DPD Free, Comb, Total
Copper
Hardness
Iron LR
Magnesium
Nitrate
Nitrite-N
Nitrite-NaNO2
Organophosphonate
Phosphate LR
Potassium
Sulfate
Turbidity
pH (Phenol Red)
Range:
0 to 0.5 mg/L
0 to 1 mg/L
0 to 10 mg/L
0 to 100 mg/L
0 to 12 mg/L
0 to 1500 mg/L
0 to 20 mg/L
0 to 200 mg/L
0 to 4 mg/L
0 to 5 mg/L
0 to 500 mg/L
0 to 6 mg/L
5 to 400 mg/L
6.8 to 8.4 mg/L
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range
15-399-29 Nitrite-N 0 to 0.5 mg/L
15-399-15 Copper 0 to 5 mg/L
15-399-30 Nitrite-NaNO2 0 to 1500 mg/L
15-399-14 Turbidity 5 to 400 mg/L
15-399-9 Chlorine-DPD Free, Comb, Total 0 to 6 mg/L
15-399-36 Potassium 0 to 12 mg/L
15-399-38 Sulfate 0 to 200 mg/L
15-399-4 Ammonia 0 to 10 mg/L
15-399-2 Alkalinity, Total 0 to 500 mg/L
15-399-7 Calcium Hardness 0 to 500 mg/L
15-399-28 Nitrate 0 to 20 mg/L
15-399-34 Phosphate LR 0 to 4 mg/L
15-399-33 pH (Phenol Red) 6.8 to 8.4 mg/L
15-399-18 Hardness 0 to 500 mg/L
15-399-31 Organophosphonate 0 to 20 mg/L
15-399-22 Iron LR 0 to 1 mg/L
15-399-25 Magnesium 0 to 100 mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 15-399-29 Supplier YSI™ Supplier No. YPM109
Add to Cart
Add to Cart

For 9300 and 9500 Photometers.

  • Includes reagents sufficient for 50 tests
TRUSTED_SUSTAINABILITY

Specifications

Type Nitrite-N
For Use With (Equipment) 9300 and 9500 Photometer
Includes Reagents, instructions, accessories
Packaging 50 Tests/Pk.
Range 0 to 0.5 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.