Learn More
Thermo Scientific™ Reagents for Free Chlorine
Description
Keep your chlorine analyzer well supplied with the reagents it needs for Free Chlorine and Total Chlorine measurement. The Chlorine XP continuously monitors a process stream using an automated DPD test for free and optionally totalchlorine. For use with the Orion™ Chlorine XP.
Specifications
Specifications
| Description | 5 Sets of DPD4 reagents for Total Chlorine |
| Type | DPD4 Reagents |
| Volume (Metric) | 600 mL |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.