Learn More
Promega RealTime-Glo Annexin V Apoptosis Assay, 1,000 assays
Catalog No. PRJA1001 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
Catalog No. Quantity
PRJA1001 1000 Assays
1 options
This item is not returnable. View return policy
Catalog No. PRJA1001 Supplier Promega Supplier No. JA1001
This item is not returnable. View return policy
A simple, non-lytic assay that allows real-time monitoring of apoptosis progression, without the need for multiple plates or complicated processing. A multimode plate reader for detecting luminescence and fluorescence is the only instrument required.
  • Simple, Plate−Based Assay for Real−Time Apoptosis Detection
  • No−wash, one−step Annexin V binding assay
  • Non−lytic. Allows continual monitoring of cell state to accurately determine apoptotic onset
  • Scalable for high−throughput screening applications
Specifications
Quantity 1000 Assays
Content And Storage -30°C to -10°C