Learn More
Gibco™ Human VEGF-165 Recombinant Protein

Description
Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water or appropriate buffered solution containing 0.1% BSA to regain full activity. Make any further dilutions of the reconstituted protein in buffered solution containing a carrier protein such as PBS with 0.1% BSA. Storage: Store the lyophilized protein at 2-8°C or -20°C for long term storage, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C. Avoid repeated freeze-thaw cycles.

Specifications
Specifications
Accession Number | P15692 |
For Use With (Application) | Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 7422 |
Molecular Weight (g/mol) | 40 kDa |
Name | Human VEGF-165 |
Purification Method | Sequential Chromatography |
Quantity | 1 mg |
Endotoxin Concentration | <0.1 ng/ μg |
Gene Alias | H-VEGF-165; MVCD1; M-VEGF-165; vascular endothelial growth factor 165 b; vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; vascular permeability factor; VEGF; VEGF164; VEGF165; Vegfa; VEGF-A; VPF |
Show More |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.