Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REC8 Polyclonal Antibody
REC8 Polyclonal Antibody
Supplier: Thermo Scientific PA598114
Description
REC8 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
REC8 | |
Polyclonal | |
Unconjugated | |
REC8 | |
cohesin Rec8p; cohesion rec8p; HR21spB; human homolog of rad21, S. pombe; kleisin-alpha; Mei8; meiosis specific sister chromatid cohesion protein; meiotic cohesion Rec8; meiotic recombination and sister chromatid cohesion phosphoprotein of the rad21p family; meiotic recombination protein REC8 homolog; meiotic recombination protein REC8-like 1; mrec; Rec8; rec8 {ECO:0000250; REC8 homolog; REC8 meiotic recombination protein; Rec8L1; REC8-like 1; Rec8p; recombination and sister chromatid cohesion protein homolog; UniProtKB:Q8C5S7} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9985 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95072 | |
REC8 | |
Recombinant Human Meiotic recombination protein REC8 homolog protein (1-270AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction