Learn More
REC8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | REC8 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6603
![]() ![]() |
Proteintech Group Inc
107931AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human REC8. This antibody can recognize the monomeric, non-phosphorylated form (63-70 kDa) of REC8 and other forms(80-100 kDa) which represent either post transla
This gene encodes a member of the kleisin family of SMC (structural maintece of chromosome) protein partners. The protein localizes to the axial elements of chromosomes during meiosis in both oocytes and spermatocytes. In the mouse, the homologous protein is a key component of the meiotic cohesion complex, which regulates sister chromatid cohesion and recombination between homologous chromosomes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene.Specifications
REC8 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O95072, Q8C5S7 | |
56739, 9985 | |
REC8 Fusion Protein Ag1250 | |
Primary | |
-20°C | |
REC8 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Cohesin Rec8p, HR21spB, REC8, REC8 homolog (yeast), REC8L1, Rec8p | |
Rec8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.