Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RecA-1 Antibody (HIS52), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064647FR
Description
RecA-1 Monoclonal specifically detects RecA-1 in Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
RecA-1 | |
Monoclonal | |
DyLight 680 | |
RecA protein, RecA1, RecA2, recaLP, Recombinase A | |
Stromal cells from rat lymph node | |
0.1 mL | |
Primary | |
Rat | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
HIS52 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
Mouse | |
Protein G purified | |
RUO | |
Recognizes RECA-1, a cell surface antigen which is expressed by all rat endothelial cells. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction