Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RECA-1 Monoclonal Antibody (HIS52)

Catalog No. PIMA180181
Change view
Click to view available options
Quantity:
100 μg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA180181 250 μg
PIMA181510 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA180181 Supplier Invitrogen™ Supplier No. MA180181
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not react with chicken, goat, guinea pig, mouse, pig, rabbit or sheep. Mouse anti Rat RECA-1 antibody, clone HIS52 recognizes RECA-1, a cell surface antigen which is expressed by all rat endothelial cells. Mouse anti Rat RECA-1 antibody, clone HIS52 recognizes RECA-1, a cell surface antigen which is expressed by all rat endothelial cells.

RECA-1 is a cell surface antigen which is expressed by all rat endothelial cells. The endothelium is the thin layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RECA-1
Applications Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone HIS52
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide
Gene Alias Reca1
Host Species Mouse
Immunogen Stromal cells from rat lymph node.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.