Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RECA-1 Monoclonal Antibody (RECA-1)
GREENER_CHOICE

Catalog No. 01676573
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-573 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-573 Supplier Invitrogen™ Supplier No. MA140240
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-40240 detects Endothelial cell antigen from rat samples. MA1-40240 has been successfully used in immunohistochemistry (frozen), immunohistochemistry (paraffin), flow cytometry and immunofluorescence applications. The MA1-40240 immunogen is cell surface antigen. Positive control: Glomerular endothelial cells, RHEC cell line; Negative control: Vascular smooth muscle cells, fibroblasts.

RECA-1 is a cell surface antigen which is expressed by all rat endothelial cells. The endothelium is the thin layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RECA-1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RECA-1
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Alias Reca1
Host Species Mouse
Immunogen Cell surface antigen.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.