Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

recA Polyclonal Antibody, Invitrogen™

Catalog No. PIPA14924
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA14924 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA14924 Supplier Thermo Scientific Supplier No. PA14924

Rabbit Polyclonal Antibody

PA1-4924 detects RecA in E. coli samples. PA1-4924 has been successfully used in Western blot, ELISA, and immunoprecipitation procedures. By western blot the antibody detects a ∽38 kDa protein (RecA) in E. coli samples. PA1-4924 immunogen corresponds to recombinant RecA protein.

E. coli RecA protein is a very important enzyme for homologous recombination and recombinational repair. Expression is induced by SOS response caused by DNA damage. The RecA protein has multiple functions, such as single stranded DNA dependent ATPase activity, DNA annealing activity, formation of D-loops and Holliday structures during homologous recombination, and co-protease activities that promote self-cleavage of the LexA repressor, lambda phage repressor and UmuD protein. RecA protein binds to single and double stranded DNA for nucleofilament formation. It carries out a central role in homologous recombination. Its is homologous to the eukaryotic Rad51 protein and Dmcl protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen recA
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and no preservative
Gene Alias RAD51; RAD51A
Gene Symbols RAD51
Host Species Rabbit
Immunogen Highly purified full-length recombinant E. coli protein.
Purification Method Affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.