Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RECK Antibody (MM0523-9G19) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | RECK |
|---|---|
| Clone | MM0523-9G19 |
| Concentration | LYOPH |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
RECK Monoclonal specifically detects RECK in Human samples. It is validated for Western Blot.Specifications
| RECK | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| O95980 | |
| 8434 | |
| IgG1 | |
| Protein G purified |
| MM0523-9G19 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| hRECK, membrane-anchored glycoprotein (metastasis and invasion), reversion-inducing-cysteine-rich protein with kazal motifs, ST15reversion-inducing cysteine-rich protein with Kazal motifs, suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein withkazal motifs), suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein withkazal motifs), Suppressor of tumorigenicity 15 protein | |
| RECK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title