Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RECK Monoclonal Antibody (F.286.6)
GREENER_CHOICE

Catalog No. PIMA514781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514781 Supplier Invitrogen™ Supplier No. MA514781
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

RECK is a cysteine-rich, extracellular protein with protease inhibitor-like domains whose expression is suppressed strongly in many tumors and cells transformed by various kinds of oncogenes. In normal cells, this membrane-anchored glycoprotein may serve as a negative regulator for matrix metalloproteinase-9, a key enzyme involved in tumor invasion and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RECK
Applications Western Blot
Classification Monoclonal
Clone F.286.6
Concentration 513 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene RECK
Gene Accession No. O95980, Q9Z0J1
Gene Alias hRECK; membrane-anchored glycoprotein (metastasis and invasion); membrane-anchored glycoprotein RECK; mRECK; RECK; reversion inducing cysteine rich protein with kazal motifs; reversion-inducing cysteine-rich protein with Kazal motifs; Reversion-inducing cysteine-rich protein with Kazal motifs precursor (mRECK); reversion-inducing-cysteine-rich protein with kazal motifs; ST15; suppression of tumorigenicity 15; suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein with kazal motifs); suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein with kazal motifs); suppressor of tumorigenicity 15 protein
Gene Symbols RECK
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Ile913 of human RECK protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313488, 53614, 8434
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.