Learn More
Novus Biologicals™ Recombinant Avian Influenza A H7N9 Hemagglutinin His Protein
Shop All R&D Systems Products

Specifications
Specifications
| Concentration | 1 mg/ml |
| For Use With (Application) | Functional Assay |
| Formulation | PBS (pH 7.4), 10% glycerol |
| Molecular Weight (g/mol) | 36 kDa |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | < 1 EU per 1ug of protein (determined by LAL method) |
| Gene Alias | H7N9HA |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Protein | Avian Influenza A H7N9 Hemagglutinin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.