Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Avian Influenza A H7N9 Hemagglutinin His Protein
SDP

Catalog No. NB151707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
0.02 mg
0.05 mg
0.1 mg
0.25 mg
0.5 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151707 0.5 mg
NB151702 0.01 mg
NB151703 0.02 mg
NB151704 0.05 mg
NB151705 0.1 mg
NB151706 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. NB151707 Supplier Novus Biologicals™ Supplier No. NBP3118500.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Specifications

Concentration 1 mg/ml
For Use With (Application) Functional Assay
Formulation PBS (pH 7.4), 10% glycerol
Molecular Weight (g/mol) 36 kDa
Quantity 0.5 mg
Storage Requirements Store at -20°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration < 1 EU per 1ug of protein (determined by LAL method)
Gene Alias H7N9HA
Purity or Quality Grade >95%, by SDS-PAGE
Protein Avian Influenza A H7N9 Hemagglutinin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.