Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Bacteria Zinc metalloprotease-1 His Protein
SDP

Catalog No. NB171730
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

∼0.5U/mg protein Active towards the generic fluorogenic substrate for matrix metalloproteinases MMP2/MMP7

Specifications

Accession Number O53649
For Use With (Application) Enzyme Activity
Formulation 10mM TRIS-HCl, pH 8.0
Molecular Weight (g/mol) M.W. Theoretical: 73.9 kDa
Quantity 50 μg
Storage Requirements Store at -20°C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias Endopeptidase, peptidase family M13, Zmp1
Research Category Immunology, Innate Immunity
Purity or Quality Grade >98%, by SDS-PAGE
Protein Zinc metalloprotease-1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.