Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Bacterial Methionine Aminopeptidase 1/METAP1 Protein
SDP

Catalog No. NBP17880310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17880310 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17880310 Supplier Novus Biologicals™ Supplier No. NBP178803100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-264 of E.coli Methionine Aminopeptidase 1/METAP1 The Recombinant E. coli Methionine Aminopeptidase 1/METAP1 Protein is derived from E. coli. The Recombinant E. coli Methionine Aminopeptidase 1/METAP1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
Formulation 20mM Tris-HCl buffer(pH 8.0) containing 10% glycerol, 2mM DTT
Gene ID (Entrez) 23173
Quantity 100μg
Storage Requirements Store at 4° short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO - research use only
Species Bacteria
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.