Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Cyno Monkey LILRB4/CD85k/ILT3 Fc Protein, CF
Click to view available options
Quantity:
50 μg
Description
Measured by its binding ability in a functional ELISA. Recombinant Cynomolgus Monkey LILRB4/CD85k/ILT3 Fc Chimera binds to Recombinant Human Apolipoprotein E3 Protein (Catalog # 4144-AE ) with a ED50 of 0.900-9.00 μg/mL.
Specifications
Specifications
Accession Number | XP_015297198.1 |
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
Gene ID (Entrez) | 11006 |
Molecular Weight (g/mol) | M.W. (Observed): 55-66 kDa, under reducing conditions., M.W. (Theroretical): 53 kDa |
Quantity | 50 μg |
Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey LILRB4/CD85k/ILT3 protein Cynomolgus LILRB-4 (Gly24-Glu259) (N-terminus) Accession # XP_015297198.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
Gene Alias | CD85 antigen-like family member K, CD85k, CD85k antigen, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5LILRB5, LIR-5subfamily B, member 4, member 4 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction