Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Lipocalin-2/NGAL His-tag Protein

Catalog No. 2357LC050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Cynomolgus Lipocalin-2/NGAL His-tag Protein is derived from CHO. The Recombinant Cynomolgus Lipocalin-2/NGAL His-tag Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number XP_005580845.2
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 3934
Molecular Weight (g/mol) 21 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived cynomolgus monkey Lipocalin-2/NGAL protein Gln21-Gly198 with C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin
Specific Reactivity Lipocalin-2/NGAL
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Buffer Supplied as a 0.2 μm filtered solution in MES and NaCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.