Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Mesothelin (aa417-693) Fc Protein
Click to view available options
Quantity:
50 μg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human CA125/MUC16 (Catalog # 5609-MU ) is immobilized at 0.5 ug/mL (100 μL/well), Recombinant Cynomolgus Monkey Mesothelin C-Terminal (aa 417-693) Fc Chimera binds with an ED50 of 4.0-20.0 ng/mL.
Specifications
Specifications
Accession Number | XP_015298164.1 |
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
Gene ID (Entrez) | 10232 |
Molecular Weight (g/mol) | M.W.-observed: 65-85 kDa, under reducing conditions, M.W.-theroretical: 58 kDa |
Quantity | 50 μg |
Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Mesothelin protein Cynomolgus Monkey Mesothelin (Asp417-Asp693) (N-terminus) Accession # XP_015298164.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Gene Alias | CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction