Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey FCAR/CD89 Protein

Catalog No. 9516FA050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Human IgA is immobilized at 5 μg/mL, 100 μL/well, the concentration of Recombinant Cynomolgus Monkey FCAR/CD89 that produces 50% of the optimal binding response is 0.250-2.50 μg/mL.

Specifications

Accession Number XP_005590398.1
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 2204
Molecular Weight (g/mol) M.W. Observed: 33-70 kDa, under reducing conditions.
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived cynomolgus monkey FCAR/CD89 protein Gln22-Asn227,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias CD89, CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
Product Type Recombinant Protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Reconstitution Reconstitute at 200μg/mL in PBS.
Buffer Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.