Learn More
Novus Biologicals Recombinant Cytomegalovirus Cytomegalovirus gH Pentamer Complex VR1814 His (C-Term) Protein
Shop All R&D Systems Products

Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Dulbecco's PBS, 0.2 um filter sterilised. |
| Quantity | 0.5 mg |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | envelope glycoprotein H |
| Purity or Quality Grade | >95% pure by SDS-PAGE |
| Protein | Cytomegalovirus gH Pentamer Complex |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.