Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant E. coli Disulfide isomerase II Protein
SDP

Catalog No. NBP26146005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP26146005 0.5 mg
NB261460100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP26146005 Supplier Novus Biologicals™ Supplier No. NBP2614600.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 21-236 of E.coli Protein disulfide isomerase II. The Recombinant E. coli Disulfide isomerase II Protein is derived from E. coli. The Recombinant E. coli Disulfide isomerase II Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-Hcl buffer (pH7.5) containing 2mM EDTA
Gene ID (Entrez) 947363
Molecular Weight (g/mol) 23.5 kDa
Quantity 0.5 mg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol dsbC
Cross Reactivity Disulfide isomerase II
Species E. coli
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.