Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant E. coli Double-Stranded Uracil-DNA Glycosylase His Protein
SDP

Catalog No. NB002850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002850 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002850 Supplier Novus Biologicals™ Supplier No. NBP27595450UG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-168 of E.coli Double-Stranded Uracil-DNA Glycosylase
  • G/U mismatch-specific DNA glycosylase, xanthine DNA glycosylase, also known as mug, belongs to the TDG/mug DNA glycosylase family.
  • It has been proposed that the Mug protein excises 3, N4-ethenocytosine and removes the uracil base from mismatches in the order of U:G>U:A, although the biological role remains unclear.
  • The enzyme Uracil-N-Glycosylase removes uracil from the DNA leaving an AP site.
  • It is capable of hydrolyzing the carbon-nitrogen bond between the sugar-phosphate backbone of the DNA and the mispaired base.
  • The complementary strand guanine functions in substrate recognition.
  • Recombinant E.coli mug protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 20% glycerol
Molecular Weight (g/mol) 21.1 kDa
Quantity 50 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias double-strand uracil-DNA glycosylase, Double-strand-specific uracil glycosylase, dsDNA specific UDG, DsUDG, EC 3.2.2.28, G/U mismatch-specific DNA glycosylase, G:T/U mismatch-specific DNA glycosylase, Mismatch-specific uracil DNA glycosylase, Mug, Xanthine DNA glycosylase
Buffer 20 mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 20% glycerol
Purity or Quality Grade >90%, by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.