Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Recombinant E. coli RNase P protein component Protein
SDP

Catalog No. NB176550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176550 250 μg
NB176548 10 μg
NB176547 100 μg
NB176552 50 μg
NB176549 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. NB176550 Supplier Novus Biologicals Supplier No. NBP312867250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

A recombinant protein corresponding to the amino acids 1-119 of E. coli rnpA

Amino Acid Sequence: MVKLAFPREL RLLTPSQFTF VFQQPQRAGT PQITILGRLN SLGHPRIGLT VAKKNVRRAH ERNRIKRLTR ESFRLRQHEL PAMDFVVVAK KGVADLDNRA LSEALEKLWR RHCRLARGS

Specifications

Accession Number NP_418159.1
Concentration 0.25 mg/ml
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 948215
Molecular Weight (g/mol) M.W. theroretical: 13.7 kDa
Quantity 250 μg
Source E. Coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <:1 EU / 1 ug of protein (determined by LAL method)
Gene Symbol rnaP
Research Category Epigenetics
Purity or Quality Grade >90%, by SDS-PAGE
Protein RNase P protein component
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.