Learn More
Novus Biologicals™ Recombinant E. coli Thioredoxin Reductase 1/TRXR1 Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 10% glycerol |
| Gene ID (Entrez) | 7296 |
| Molecular Weight (g/mol) | 34.6kDa |
| Name | Thioredoxin Reductase 1/TRXR1 Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Immunogen | MGTTKHSKLL ILGSGPAGYT AAVYAARANL QPVLITGMEK GGQLTTTTEV ENWPGDPNDL TGPLLMERMH EHATKFETEI IFDHINKVDL QNRPFRLNGD NGEYTCDALI IATGASARYL GLPSEEAFKG RGVSACATCD GFFYRNQKVA VIGGGNTAVE EALYLSNIAS EVHLIHRRDG FRAEKILIKR LMDKVENGNI ILHTNRTLEE VTGDQMGVTG VRLRDTQNSD NIESLDVAGL FVAIGHSPNT AIFEGQLELE NGYIKVQSGI HGNATQTSIP GVFAAGDVMD HIYRQAITSA GTGCMAALDA ERYLDGLADA K |
| Storage Requirements | -80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Bacteria |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.