Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant eGFP His (N-Term) His (C-Term) Protein
SDP

Catalog No. NBP199915 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP199915 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP199915 Supplier Novus Biologicals™ Supplier No. NBP199915
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: In vitro assay, In vivo assay, SDS-Page

A recombinant protein with a N-Terminal and a C-terminal His-tag and corresponding to eGFP The Recombinant eGFP His (N-Term) His (C-Term) Protein is derived from E. coli. The Recombinant eGFP His (N-Term) His (C-Term) Protein has been validated for the following applications: In vitro assay, In vivo assay, SDS-Page.

Specifications

Formulation Ready to use liquid in PBS with no additives
Name GFP Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Purity or Quality Grade >97%, by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.