Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Guinea Pig IL-1 beta/IL-1F2 Protein

Catalog No. p-4718975
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in MES, NaCl, DTT, and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in MES, NaCl, DTT, and EDTA.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Guinea Pig IL-1 beta/IL-1F2 Protein is derived from E. coli. The Recombinant Guinea Pig IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in MES, NaCl, DTT, and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 100135556
Molecular Weight (g/mol) 17.4 kDa
Name IL-1 beta/IL-1F2
Quantity 10 μg
Source E. coli-derived guinea pig IL-1 beta/IL-1F2 protein Thr115-Ser266,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Monomer
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.