Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Hepatitis C Virus His Protein
SDP

Catalog No. NBP141149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141149 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141149 Supplier Novus Biologicals™ Supplier No. NBP141149
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1225-1456 of HCV Hepatitis C Virus The Recombinant Virus Hepatitis C Virus Protein is derived from E. coli. The Recombinant Virus Hepatitis C Virus Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT
Molecular Weight (g/mol) 27.9kDa
Name Hepatitis C Virus Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSVAHL HAPTGSGKST KVPAAYAAQG YKVLVLNPSV AATLGFGAYM SKAHGVDPNI RTGVRTITTG SPITYSTYGK FLADGGCSGG AYDIIICDEC HSTDATSILG IGTVLDQAET AGARLVVLAT ATPPGSVTVS HPNIEEVALS TTGEIPFYGK AIPLEVIKGG RHLIFCHSKK KCDELAAKLV ALGINAVAYY RGLDVSVIPT SGDVVVVSTD ALMTGFTGDF DSVIDCNT
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.